missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IFN-γ Recombinant Human Protein DFS Item

Recombinant Human Interferon-γ (IFN-γ) is a bioactive protein intended for use in cell culture applications

$282.00 - $1,358.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias IFN-γ
View More Specs

Disclaimers:

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC4031 Gibco™
LSPHC4031
100μg Each for $282.00
PHC4033 Gibco™
LSPHC4033
1mg Each for $1,358.00
Specifications

Description

IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone resorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytostasis of human WiDr cells
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Dry Ice.

Description & Specifications

Specifications

95%
<0.1ng/μg
IFN-γ
Sequential Chromatography
Liquid
3458