missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Hybridoma-SFM
Specifically developed to support the growth of hybridomas for antibody production
Supplier: Gibco™ 12045084
Description
- Hybridoma-SFM is a serum-free, very low-protein (<20 μg/mL) medium specifically developed to support the growth of hybridomas for antibody production
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Hybridoma-SFM is a complete medium that is ready to use, no supplementation required
- Store at 2°C to 8°C, protected from light
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Low Protein, Serum-free | |
Serum Free Medium (SFM) | |
Glutamine, Phenol Red | |
Gibco™ | |
Room Temperature |
Hybridomas | |
Liquid | |
Serum-free | |
Sustainable packaging | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.