missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ GM-CSF Recombinant Swine Protein DFS Item

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPSC2015

 View more versions of this product

Catalog No. PSC2015



Description

Description

Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture
< 0.1 ng/µg
Gibco®
Approved for shipment on Wet or Dry Ice
Lyophilized
Recombinant, Ligand
Signal Transduction, Stem Cell Research, Drug Development, Oncology
397208
Sequential Chromatography
25μg
Store in refrigerator (2–8°C).
25 µg
Carrier-Free
E. coli
Cytokines & Receptors
GM-CSF, G-CSF, & M-CSF
Swine⁄Porcine
14 kDa
95 %
GM-CSF
Documents

For Research Use Only. Not for use in diagnostic procedures.