missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ GM-CSF Recombinant Swine Protein DFS Item

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPSC2014

 View more versions of this product

Catalog No. PSC2014


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; GM-CSF Recombinant Swine Protein; 10ug; Source: Swine/Porcine; Purity: 95%

Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture
397208
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
14kDa
95%
Swine/Porcine
Carrier Protein
GM-CSF, G-CSF, & M-CSF
GM-CSF
Cytokines & Receptors