Please login to your online account to display your discounted pricing

Gibco™ GM-CSF Recombinant Swine Protein DFS Item


Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPSC2011

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PSC2011

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Molecular Weight 14kDa
Quantity 100μg
Purification Method Sequential Chromatography
Purity 95%
Source Swine/Porcine
Format Lyophilized
Structural Form Recombinant/Ligand
Without Additives Carrier Protein
Gene Alias GM-CSF
Protein Subtype GM-CSF, G-CSF, & M-CSF
Expression System E. coli
Endotoxin Concentration <0.1ng/μg
For Use With (Application) Cell Culture; Mammalian Cell Culture

Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.