missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ GM-CSF Recombinant Rat Protein DFS Item

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

$280.00 - $1,266.00

Specifications

Purity 95%
Purification Method Exchange Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias GM-CSF
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRC2011 Gibco™
LSPRC2011
100μg Each for $1,266.00
PRC2015 Gibco™
LSPRC2015
10μg Each for $280.00
PRC2016 Gibco™
LSPRC2016
25μg Each for $355.00
Specifications

Description

Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
GM-CSF
Exchange Chromatography
Lyophilized
116630