Please login to your online account to display your discounted pricing

Gibco™ GM-CSF Recombinant Rat Protein DFS Item


Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC2011

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC2011

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 14.6kDa
Purification Method Exchange Chromatography
Purity 95%
Quantity 100μg
Source Rat
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype GM-CSF, G-CSF, & M-CSF
Endotoxin Concentration <0.1ng/μg
Gene Alias GM-CSF
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 116630

Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.