Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder DFS Item
Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Manufacturer:  Gibco™ 11811023
 View more versions of this product
Catalog No. 11811023
Description
Description
Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3) – I and APH (3) – II resistance genes.
  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as powder and should be made into a stock solution of 10 – 50mg/mL in water
Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation
Specifications
Specifications
Selective Antibiotic
Room Temperature
Geneticin™
24 Months
Antibiotic (Selective)
Geneticin™ (G-418)
Eukaryotic Selection⁄Stable Cell Line Generation
Powder
1g
Eukaryotic Cell, Prokaryotic Cell
SDS
Documents
Product Certifications
Promotions
Promotions
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.