missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ GDNF Recombinant Human Protein DFS Item

Glial-Derived Neurotrophic Factor (GDNF) is a bioactive protein intended for use in cell culture applications

$244.00 - $1,380.00

Specifications

Purity 98%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias G-DNF
View More Specs

Disclaimers:

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC7044 Gibco™
LSPHC7044
5μg Each for $244.00
PHC7041 Gibco™
LSPHC7041
100μg Each for $1,380.00
PHC7045 Gibco™
LSPHC7045
10μg Each for $378.00
Specifications

Description

GDNF is mainly known for its affect on neurons, but it is also involved in other biological processes including kidney development.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by inducing neurite outgrowth on PC-12 cells in the presence of recombinant soluble GFR-α
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

98%
<0.1ng/μg
G-DNF
Sequential Chromatography
Lyophilized
2668