Please login to your online account to display your discounted pricing

Gibco™ G-CSF Recombinant Mouse Protein DFS Item

Recombinant G-CSF is a bioactive protein intended for use in cell culture applications

$376.00 - $1,350.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias G-CSF
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PMC2031

DFS Item
 
gibco™
LSPMC2031
100μg Each for $1,350.00

PMC2034

DFS Item
 
gibco™
LSPMC2034
10μg Each for $376.00

PMC2035

DFS Item
 
gibco™
LSPMC2035
25μg Each for $726.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias G-CSF

G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.