Please login to your online account to display your discounted pricing

Gibco™ G-CSF Recombinant Mouse Protein DFS Item


Recombinant G-CSF is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC2034

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC2034

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
Molecular Weight 19kDa
Protein Family Cytokines & Receptors
Protein Subtype GM-CSF, G-CSF, & M-CSF
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Quantity 10μg
Purity 95%
Format Lyophilized
Gene ID (Entrez) 12985
Expression System E. coli
Structural Form Recombinant/Ligand
Validated Application Cell Culture
Without Additives Carrier Protein
Source Mouse
Storage Requirements Store at 2° to 8°C.
Gene Alias G-CSF

G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.