Please login to your online account to display your discounted pricing

Gibco™ G-CSF Recombinant Human Protein DFS Item


Recombinant Human Granulocyte Colony-Stimulating Factor (G-CSF) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC2033

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHC2033

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Proliferation; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
Molecular Weight 19kDa
Purity 95%
Quantity 1mg
Source Human
Storage Requirements Lyophilized hG-CSF should be stored at 2° to 8°C, preferably desiccated. Store reconstituted hG-CSF at ≤ -20°C. Avoid freeze-thaw cycles.
Without Additives Carrier Protein
Structural Form Mutant/Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Gene Alias G-CSF
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
Format Lyophilized
Gene ID (Entrez) 1440
Purification Method Sequential Chromatography
Protein Subtype GM-CSF, G-CSF, & M-CSF

G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse NFS-60 cells
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.