Please login to your online account to display your discounted pricing

Freestyle™ CHO Expression Medium DFS Item


Animal origin-free, chemically defined, protein-free medium

Manufacturer: life technologies  LS12651022

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12651022

  • / Each
 

Description & Specifications

Specifications

Quantity 6 x 1000mL
Storage Requirements Storage conditions: 2°C to 8°C. Protect from light. Shelf life: 12 months from date of manufacture
Cell Line CHO, FreeStyle™ CHO-S Cells
Format Liquid
For Use With (Application) Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Additive No glutamine, no phenol red