missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ FGF-Basic (aa 1-155) Recombinant Human Protein DFS Item

FGF-Basic (AA 1-155) is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHG0264

 View more versions of this product

Catalog No. PHG0264



Description

Description

Fibroblast growth factor-basic is known as FGF-basic, bFGF, FGF 2, FGF-β, and heparin binding growth factor. FGF-basic is involved in a number of biological processes, including embryonic development and differentiation, neuron differentiation, survival, regeneration, and the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: proteins bioassayed with Gibco media
  • High-purity, recombinant, bioactive protein: to help ensure growth factors from Life Technologies are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

17.2kDa
<0.1ng/μg
Sequential Chromatography
Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Differentiation & Development
Growth Factors & Receptors
Store at 2° to 8°C.
Recombinant/Ligand
E. coli
95%
2247
10μg
Human
FGFs (Fibroblast Growth Factors)
FGF-Basic (AA 1-155)
Cell Culture
Carrier Protein
Documents

For Research Use Only. Not for use in diagnostic procedures.