Please login to your online account to display your discounted pricing

Gibco™ FGF-8b Recombinant Human Protein DFS Item

Recombinant Human Fibroblast Growth Factor 8B (FGF-8B) is a bioactive protein intended for use in cell culture applications

$247.00 - $4,465.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias FGF-8b
View More Specs

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHG0271

DFS Item
 
gibco™
LSPHG0271
100μg Each for $998.00

PHG0273

DFS Item
 
gibco™
LSPHG0273
1mg Each for $4,465.00

PHG0274

DFS Item
 
gibco™
LSPHG0274
10μg Each for $247.00

PHG0276

DFS Item
 
gibco™
LSPHG0276
25μg Each for $311.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias FGF-8b

FGF-8b is one of the isoforms of FGF-8, and is involved in many biological processes including angiogenesis.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.