Please login to your online account to display your discounted pricing

Gibco™ FGF-8b Recombinant Human Protein DFS Item


Recombinant Human Fibroblast Growth Factor 8B (FGF-8B) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHG0276

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHG0276

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22.5kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 25μg
Source Human
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Truncated/Recombinant/Ligand
Protein Subtype FGFs (Fibroblast Growth Factors)
Endotoxin Concentration <0.1ng/μg
Gene Alias FGF-8b
Validated Application Cell Culture
Protein Family Growth Factors & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Differentiation & Development
Format Lyophilized

FGF-8b is one of the isoforms of FGF-8, and is involved in many biological processes including angiogenesis.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.