missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, ultra-low IgG, US Origin DFS Item

US-qualified performance sera for general cell culture with common cell lines

$1,262.00 - $1,262.00

Specifications

Format Bottle
Endotoxin Concentration ≤50 EU/mL (levels routinely ≤ 10 EU/mL)
Heat Inactivated No
Hemoglobin Concentration ≤25 mg/dL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
16250078 Life Technologies
LS16250078
500mL Each for $1,262.00
Specifications

Description

  • Sera Category: Specialty
  • Origin: United States
  • IgG levels are less than 5μg/mL
  • BVD titer is low or not detectable
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25mg/dL

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

Bottle
≤50 EU/mL (levels routinely ≤ 10 EU/mL)
No
≤25 mg/dL