Please login to your online account to display your discounted pricing

Fetal Bovine Serum, Qualified, USDA-approved Regions DFS Item


Trusted quality for consistent performance

Manufacturer: life technologies  LS10437010

 View more versions of this product

Catalog No. 10437010

  • / Each
 

Description & Specifications

Specifications

Grade Qualified
Format Bottle
Hemoglobin Concentration ≤25mg/dL
Quantity 100mL
Endotoxin Concentration ≤50 EU/mL
Heat Inactivated No