Please login to your online account to display your discounted pricing

Fetal Bovine Serum, Qualified, Heat Inactivated, USDA-approved Regions DFS Item


General stem cell culture media and reagents; sera for specialty research and specific assays

Manufacturer: life technologies  LS10438018

 View more versions of this product

 Disclaimers

For In Vitro Diagnostic Use.

Catalog No. 10438018

  • / Each
 

Description & Specifications

Specifications

Quantity 100mL
Heat Inactivated Yes
Endotoxin Concentration ≤ 50 EU/mL
Format Bottle
Hemoglobin Concentration ≤25 mg/dL
Grade Qualified