Please login to your online account to display your discounted pricing

Fetal Bovine Serum, Qualified, Heat Inactivated, Australia Origin DFS Item


Standard sera for robust cell lines

Manufacturer: life technologies  LS10100154

 View more versions of this product

Catalog No. 10100154

  • / Each
 

Description & Specifications

Specifications

Heat Inactivated Yes
Quantity 1000mL
Format Bottle
Hemoglobin Concentration ≤30 mg/dL (levels routinely ≤25 mg/dL)
Endotoxin Concentration ≤ 10 EU/mL
Grade Qualified