missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Qualified, Heat Inactivated, Australia Origin DFS Item

Standard sera for robust cell lines

$350.00 - $2,540.00

Specifications

Format Bottle
Grade Qualified
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤30 mg/dL (levels routinely ≤25 mg/dL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10100154 Life Technologies
LS10100154
1000mL Each for $2,540.00
10100139 Life Technologies
LS10100139
100mL Each for $350.00
10100147 Life Technologies
LS10100147
500mL Each for $1,312.00
Specifications

Description

  • Sera Category: Secure
  • Origin: Australia
  • Endotoxin level: ≤10EU/mL
  • Hemoglobin level: ≤30mg/dL (levels routinely ≤25mg/dL)

Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

Bottle
≤ 10 EU/mL
≤30 mg/dL (levels routinely ≤25 mg/dL)
Qualified
Yes