Please login to your online account to display your discounted pricing

Fetal Bovine Serum, Qualified, Australia Origin DFS Item


Standard sera for robust cell lines

Manufacturer: life technologies  LS10099141

 View more versions of this product

Catalog No. 10099141

  • / Each
 

Description & Specifications

Specifications

Hemoglobin Concentration ≤30 mg/dL (levels routinely ≤25 mg/dL)
Endotoxin Concentration ≤ 10 EU/mL
Quantity 500mL
Grade Qualified
Format Bottle
Heat Inactivated No