Please login to your online account to display your discounted pricing

Fetal Bovine Serum, mesenchymal stem cell-Qualified, USDA-approved Regions, Standard (Sterile-Filtered) DFS Item


US certified performance plus sera for broad range of cell types, especially sensitive cell lines

Manufacturer: life technologies  LS12662011

 View more versions of this product

Catalog No. 12662011

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance USDA-Approved
Description Fetal Bovine Serum Mesenchymal Stem Cell-Qualified, USDA-Approved, 100mL, Liquid, ≤50 EU/mL Endotoxin Concentration, ≤25mg/dL Hemoglobin Concentration, –5 to –20°C Storage Temperature, Fetal, Cattle/Bovine Species, Frozen
Additional Assays Basic cell culture, specialty research, and specific assays
Endotoxin Concentration ≤50 EU/mL (levels routinely ≤10 EU/mL)
Heat Inactivated No
Grade MS Cell-Qualified
Formulation Liquid
Format Bottle
For Use With (Application) Basic cell culture, specialty research, and specific assays
Hemoglobin Concentration ≤25 mg/dL
Manufacturing Origin USDA-Approved
Origin Countries meeting USDA importation requirements
Size 100mL
Storage –5 to –20°C
Quantity 100mL
Storage Requirements –5 to –20°C
USDA Safety Testing Included Yes
Product Type Fetal Bovine Serum Mesenchymal Atem Aell-Qualified
Tested For Ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells
Type Fetal Bovine Serum Mesenchymal Atem Aell-Qualified
Purity MS Cell-qualified