Please login to your online account to display your discounted pricing

Fetal Bovine Serum, embryonic stem cell-Qualified, USDA-approved Regions DFS Item


US certified performance plus sera for broad range of cell types, especially sensitive cell lines

Manufacturer: life technologies  LS10439016

 View more versions of this product

Catalog No. 10439016

  • / Each
 

Description & Specifications

Specifications

Format Bottle
Quantity 100mL
Endotoxin Concentration ≤ 50 EU/mL
Grade ES Cell-Qualified
Hemoglobin Concentration ≤25 mg/dL
Heat Inactivated No