Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, dialyzed , US Origin DFS Item

Use for general cell culture with common cell lines

$343.00 - $1,270.00

Specifications

Format Bottle
Endotoxin Concentration ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
Heat Inactivated No
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

26400036

DFS Item
 
life technologies
LS26400036
100mL Each for $343.00

26400044

DFS Item
 
life technologies
LS26400044
500mL Each for $1,270.00
Description & Specifications

Specifications

Format Bottle
Endotoxin Concentration ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
Heat Inactivated No

  • Sera Category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

Cell Culture, Mammalian Cell Culture