Please login to your online account to display your discounted pricing

Fetal Bovine Serum, charcoal stripped, USDA-approved Regions DFS Item


Secure sera for cell culture with least viral risk sera from BSE-free regions

Manufacturer: life technologies  LS12676011

 View more versions of this product

 Disclaimers

For In Vitro Diagnostic Use.

Catalog No. 12676011

  • / Each
 

Description & Specifications

Specifications

Quantity 50mL
Heat Inactivated No
Grade Research Grade
Format Bottle