Please login to your online account to display your discounted pricing

Fetal Bovine Serum, Certified, Heat Inactivated, US Origin DFS Item


Secure sera for cell culture with least viral risk sera from BSE-free regions

Manufacturer: life technologies  LS10082139

 View more versions of this product

 Disclaimers

For In Vitro Diagnostic Use.

Catalog No. 10082139

  • / Each
 

Description & Specifications

Specifications

Source Fetal Bovine
Heat Inactivated Yes
Grade Certified
Hemoglobin Concentration ≤10 mg/dL
Product Type FBS
Quantity 100mL
Endotoxin Concentration ≤5 EU/mL
Format Bottle