Please login to your online account to display your discounted pricing

Gibco™ EPO Recombinant Human Protein DFS Item

Erythropoietin (EPO) is a bioactive protein intended for use in cell culture applications

$386.00 - $1,846.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias EPO, Erythropoietin
View More Specs

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC2054

DFS Item
 
gibco™
LSPHC2054
500 IU Each for $532.00

PHC9631

DFS Item
 
gibco™
LSPHC9631
100μg Each for $1,846.00

PHC9634

DFS Item
 
gibco™
LSPHC9634
10μg Each for $386.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias EPO, Erythropoietin

Erythropoietin is a member of the EPO/TPO family. It regulates red cell production by promoting erythroid differentiation and initiating hemoglobin synthesis. This protein also has neuroprotective activity against a variety of potential brain injuries and antiapoptotic functions in several tissue types.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by in vivo bioassay measuring dose-dependent increase in reticulocyte count with normocythaemic mice
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.