missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Eotaxin Recombinant Human Protein DFS Item

Recombinant human eotaxin is a bioactive protein intended for use in cell culture applications

$1,508.00 - $4,105.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Eotaxin
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC1431 Gibco™
LSPHC1431
100μg Each for $1,508.00
PHC1433 Gibco™
LSPHC1433
1mg Each for $4,105.00
Specifications

Description

Eotaxin is a member of the PF-4 family of chemokines. Eotaxin plays a role in inducing calcium flux in eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
Eotaxin
Sequential Chromatography
Lyophilized
6356