missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Eotaxin Recombinant Human Protein DFS Item

Recombinant human eotaxin is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC1433

 View more versions of this product

Catalog No. PHC1433


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; Eotaxin Recombinant Human Protein; 1mg; Source: Human; Purity: 95%

Eotaxin is a member of the PF-4 family of chemokines. Eotaxin plays a role in inducing calcium flux in eosinophils.

Shipping Condition: Wet Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture
6356
Sequential Chromatography
1mg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
∽8kDa
95%
Human
Carrier Protein
Eotaxins
Eotaxin
Chemokines & Receptors