Please login to your online account to display your discounted pricing

DYNAL™ Dynabeads™ Mouse T-Activator CD3/CD28 for T-Cell Expansion and Activation, Monoclonal DFS Item


4.5μm superparamagnetic beads with anti-CD3 and anti-CD28 antibodies covalently bound to the bead surface

Manufacturer: life technologies  LS11456D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11456D

  • / Each
 

Description & Specifications

Specifications

Quantity 0.4mL
Sample Type Lymph Node, Spleen
Applications Cell Activation and Expansion, Cell Analysis, Cell Isolation and Expansion, Mouse Cell Activation and Expansion
Cross Reactivity Mouse
Storage Requirements Storage: 2°C to 8°C.
Includes 0.4mL Dynabeads Mouse T-Activator CD3/CD28 is supplied as a suspension containing 4 x 107 Dynabeads/mL in phosphate buffered saline (PBS), pH 7.4, with 0.1% bovine serum albumin (BSA)
Purification Method Dynabeads Untouched or Dynabeads FlowComp
Purity Research Grade