Please login to your online account to display your discounted pricing

DYNAL™ Dynabeads™ Human T-Activator CD3/CD28 for T Cell Expansion and Activation, Monoclonal DFS Item


For activation and expansion of human T cells

Manufacturer: life technologies  LS11131D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11131D

  • / Each
 

Description & Specifications

Specifications

Includes Dynabeads Human T-Activator CD3/CD28 are supplied as a suspension containing 4 x 107 Dynabeads/mL in phosphate buffered saline (PBS), pH 7.4, with 0.1% human serum albumin (HSA).
Applications Cell Activation and Expansion, Cell Analysis, Cell Isolation, Cell Isolation and Expansion, Human Cell Activation and Expansion, Human Cell Isolation
Purity Research Grade
Quantity 2mL
Sample Type Cell Cultures, PBMC
Storage Requirements Storage: 2°C to 8°C. Do not freeze.
Cross Reactivity Human