Please login to your online account to display your discounted pricing

DYNAL™ Dynabeads™ Human T-Activator CD3/CD28 for T Cell Expansion and Activation, Monoclonal DFS Item


For activation and expansion of human T cells

Manufacturer: life technologies  LS11161D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11161D

  • / Each
 

Description & Specifications

Specifications

Includes The product contains 0.4mL Dynabeads Human T-Activator CD3/CD28 in phosphate buffered saline (PBS), pH 7.4, containing 0.1% human serum albumin (HSA). Store at 2-8°C. 4 x 107 beads/mL suspension.
Applications Cell Activation and Expansion, Cell Analysis, Cell Isolation and Expansion, Human Cell Activation and Expansion
Purification Method Dynabeads Untouched or Dynabeads FlowComp
Purity Research Grade
Quantity 0.4mL
Storage Requirements Storage: 2°C to 8°C.
Cross Reactivity Human