Please login to your online account to display your discounted pricing

DYNAL™ Dynabeads™ Human T-Activator CD3/CD28/CD137 DFS Item


Coupled with anti-CD3, anti-CD28, and anti-CD137 antibodies

Manufacturer: life technologies  LS11163D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11163D

  • / Each
 

Description & Specifications

Specifications

Includes 2mL Dynabeads Human T-Activator CD3/CD28/CD137 is supplied as a suspension containing 5 x 105 Dynabeads/mL in phosphate buffered saline (PBS), pH 7.4, with 0.1% human serum albumin (HSA)
Applications Cell Activation and Expansion, Cell Analysis, Cell Isolation and Expansion, Human Cell Activation and Expansion
Quantity 2mL
Storage Requirements Storage: 2°C to 8°C.
Cross Reactivity Human