missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ DMEM, high glucose, no glutamine DFS Item

Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, No Glutamine is used in cell culture processing applications.

Manufacturer:  Gibco™ LS11960077

 View more versions of this product

Catalog No. 11960077



Requires : Supplementation

Description

Description

  • Contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium
  • Originally formulated with low glucose (1g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (3.7g/L), and therefore requires a 5 to 10% CO2 environment to maintain physiological pH
  • Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, cell lines such as HeLa, 293, Cos-7, and PC-12
Specifications

Specifications

Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
No HEPES
6 x 1000mL
Room Temperature
1000 ml
No Glutamine
High Glucose
Gibco®
Standard Serum Supplementation
No Sodium Pyruvate
Liquid
Phenol Red
SDS
Documents

When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For human ex vivo tissue and cell culture processing applications.