missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ DMEM, high glucose, no glutamine
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, No Glutamine is used in cell culture processing applications.
Manufacturer: Gibco™ LS11960069
Requires : Supplementation
Description
- Contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium
- Originally formulated with low glucose (1g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate
- Contains no proteins, lipids, or growth factors
- Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
- Uses sodium bicarbonate buffer system (3.7g/L), and therefore requires a 5 to 10% CO2 environment to maintain physiological pH
- Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, cell lines such as HeLa, 293, Cos-7, and PC-12
Specifications
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
No HEPES | |
10 x 500mL | |
Room Temperature | |
500 ml | |
No Glutamine |
High Glucose | |
Gibco® | |
Standard Serum Supplementation | |
No Sodium Pyruvate | |
Liquid | |
Phenol Red |
When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For human ex vivo tissue and cell culture processing applications.