Please login to your online account to display your discounted pricing

Gibco™ DMEM, High Glucose, HEPES DFS Item


Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, HEPES is used in cell culture processing applications.

Manufacturer: gibco™  LS12430112

 View more versions of this product

Supplementation

 Disclaimers

When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco™ DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For human ex vivo tissue and cell culture processing applications.

Catalog No. 12430112

  • / Each
 

Description & Specifications

Specifications

Product Type DMEM media
Shelf Life 12 months from the date of manufacture
Storage Requirements Storage: 2-8° C. Protect from light
With Additives High Glucose, L-glutamine, HEPES, Phenol Red
Without Additives Sodium Pyruvate
For Use With (Application) Antibody and cell culture production, Mammalian cell culture
Formulation Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D-Calcium pantothenate, D-Glucose (Dextrose), Ferric Nitrate (Fe(NO3)3·9H2O), Folic Acid, Glycine, HEPES, L-Arginine hydrochloride, L-Cystine 2HCl, L-Glutamine, L-Histidine hydrochloride-H2O, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine disodium salt dihydrate, L-Valine, Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Phenol Red, Potassium Chloride (KCl), Pyridoxine hydrochloride, Riboflavin, Sodium Bicarbonate (NaHCO3), Sodium Chloride (NaCl), Sodium Phosphate monobasic (NaH2PO4-H2O), Thiamine hydrochloride, i-Inositol
Quantity 5L
Format Liquid
Quantity 5L
With Additives No Sodium Pyruvate; High Glucose; L-Glutamine; Phenol Red

  • Contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium
  • Originally formulated with low glucose (1g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (3.7g/L), and therefore requires a 5 to 10% CO2 environment to maintain physiological pH
  • Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, cell lines such as HeLa, 293, Cos-7, and PC-12

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.