missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Human Dkk-1 Recombinant Protein DFS Item

Recombinant Human Dickkopf Homolog 1 (DKK1) is a bioactive protein intended for use in cell culture applications

$401.00 - $9750.00

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 42 kDa
Purification Method Sequential Chromatography
Gene Alias DKK-1
Biological Activity Linear range of 0.3 to 40 ng⁄ml
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC9214 DFS Item
View Documents Promotion Details
Gibco™
PHC9214
10μg
Each for $401.00
 
PHC9215 DFS Item
View Documents Promotion Details
Gibco™
PHC9215
25μg
Each for $1,114.00
 
PHC9211 DFS Item
View Documents Promotion Details
Gibco™
PHC9211
100μg
Each for $3,525.00
 
PHC9213 DFS Item
View Documents Promotion Details
Gibco™
PHC9213
1mg
Each for $9,750.00
 
Description

Description

Human Dkk-1 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

DKK1 is a member of the dickkopf family. It is a secreted protein with two cysteine-rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by a functional ELISA measuring the ability of human DKK1 to bind immobilized recombinant human LRP6/Fc chimera receptor
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
Sequential Chromatography
Linear range of 0.3 to 40 ng⁄ml
Store lyophilized human DKK1 at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ -20°C. Avoid repeated freeze⁄thaw cycles.
< 0.1 ng/μg
Wet Ice
Mammalian
Recombinant, Ligand
Other Proteins
Gibco™
42 kDa
DKK-1
Human
Carrier-Free
Human
Linear range of 0.3 to 40 ng⁄ml
Cytokines & Receptors
Lyophilized
Oncology, Signal Transduction
>95% by SDS-PAGE
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.