missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ CXCL1 (KC) Recombinant Mouse Protein DFS Item

Recombinant CXCL1 (KC) is a bioactive protein intended for use in cell culture applications

$947.10 - $947.10

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias CXCL1, KC
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PMC1061 Gibco™
LSPMC1061
100μg Each for $947.10
Specifications

Description

CXCL1 is a chemokine that induces the infiltration of neutrophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
CXCL1, KC
Sequential Chromatography
Lyophilized
14825