Please login to your online account to display your discounted pricing

Gibco™ CXCL1 (KC) Recombinant Mouse Protein DFS Item


Recombinant CXCL1 (KC) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC1061

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC1061

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8kDa
Quantity 100μg
Storage Requirements Store at 2° to 8°C.
Purification Method Sequential Chromatography
Purity 95%
Source Mouse
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype GRO/KC/CINC Chemokines
Endotoxin Concentration <0.1ng/μg
Gene Alias CXCL1, KC
Validated Application Cell Culture
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 14825

CXCL1 is a chemokine that induces the infiltration of neutrophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.