Please login to your online account to display your discounted pricing

Gibco™ CXCL1 (GROα) Recombinant Human Protein DFS Item


Recombinant Human Chemokine (C-X-C motif) ligand 1 (Also known as CXCL1, GROα, melanoma growth stimulating activity, MSGA, Neutrophil-activating protein 3, NAP-3) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1063

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHC1063

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8kDa
Purity 95%
Quantity 1mg
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Expression System E. coli
Format Lyophilized
Purification Method Sequential Chromatography
Source Human
Storage Requirements Store at 2° to 8°C.
Protein Subtype GRO/KC/CINC Chemokines
Endotoxin Concentration <0.1ng/μg
Gene Alias CXCL1, GROα
Validated Application Cell Culture
Protein Family Chemokines & Receptors
For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene ID (Entrez) 2919

CXCL1 is a chemokine that induces the infiltration of neutrophils.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.