Please login to your online account to display your discounted pricing

Gibco™ Cripto Recombinant Human Protein DFS Item

Recombinant human Cripto is a bioactive protein intended for use in cell culture applications.

$362.25 - $1,243.20

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Cripto
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC8011

DFS Item
 
gibco™
PHC8011
100μg Each for $1,243.20

PHC8014

DFS Item
 
gibco™
PHC8014
10μg Each for $362.25
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Cripto
Gene ID (Entrez) 4507424, 57932

Cripto is a member of the EGF-related family of growth factors.

Shipping Condition: Wet Ice.