Please login to your online account to display your discounted pricing

Gibco™ Cripto Recombinant Human Protein DFS Item


Recombinant human Cripto is a bioactive protein intended for use in cell culture applications.

Manufacturer: gibco™  LSPHC8011

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC8011

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
Molecular Weight 17.8kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 100μg
Source Human
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype EGF (Epidermal Growth Factor)
Endotoxin Concentration <0.1ng/μg
Gene Alias Cripto
Validated Application Cell Culture
Protein Family Growth Factors & Receptors
Expression System Mammalian
Format Lyophilized
Gene ID (Entrez) 4507424, 57932

Cripto is a member of the EGF-related family of growth factors.

Shipping Condition: Wet Ice.