Please login to your online account to display your discounted pricing

Hybridoma AGT™ Medium DFS Item


Animal origin-free, chemically defined, protein-free medium

Manufacturer: life technologies  LS12372025

 View more versions of this product

 Disclaimers

For research use/further cell culture manufacturing. Not for use in diagnostic procedures.

Catalog No. 12372025

  • / Each
 

Description & Specifications

Specifications

Quantity 1L
For Use With (Application) Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Format Powder (AGT)
Species Mouse
Additive No glutamine, no phenol red
Storage Requirements Storage conditions: 2-8°C, dark and dry. Shelf life: 24 months from date of manufacture