missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ CD Hybridoma AGT™ Medium, Nonsterile, Sodium Bicarbonate Buffer DFS Item

Animal origin-free, chemically defined, protein-free medium

$4,720.00 - $4,720.00

Specifications

Additive No Glutamine, no phenol red, no sodium pyruvate, low glucose
Storage Requirements Storage conditions: 2° to 8°C, dark and dry. Shelf life: 24 months from date of manufacture
View More Specs

Disclaimers:

For Research Use Only or Further Manufacturing. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12372003 Life Technologies
LS12372003
10 kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12372001 Life Technologies
LS12372001
100L Each for $4,720.00
Specifications

Description

Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Formulated without L-glutamine for stability
  • Animal origin-free, protein-free and chemically defined formulation
  • Convenient, easy-to-use Advanced Granulation Technology (AGT™) format

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet ice

Description & Specifications

Specifications

No Glutamine, no phenol red, no sodium pyruvate, low glucose
Storage conditions: 2° to 8°C, dark and dry. Shelf life: 24 months from date of manufacture