missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bovine Serum, New Zealand origin
US-qualified performance sera for general cell culture with common cell lines
$56.00 - $337.00
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
|---|---|
| Species | Cattle/Bovine |
| Age | 12 to 24 months |
| Country of Origin | New Zealand |
| Form | Liquid |
Description
- Sera Category: Secure
- Origin: New Zealand
- Collected from cattle of age 12-24 months
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
| Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
| 12 to 24 months | |
| Liquid | |
| Adult Bovine Serum | |
| Sterile-filtered | |
| Gibco™ |
| Cattle/Bovine | |
| New Zealand | |
| Bottle | |
| Sterile-filtered | |
| Sterile-filtered | |
| Frozen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.