missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Bovine Serum, New Zealand origin DFS Item

US-qualified performance sera for general cell culture with common cell lines

Supplier:  Gibco™ 16170086

 View more versions of this product

Catalog No. 16170086



Description

Description

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from cattle of age 12-24 months

Cell Culture, Mammalian Cell Culture

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
New Zealand
Bottle
100 mL
Cattle/Bovine
Sustainable packaging
Frozen
12 to 24 Months
Liquid
Adult Bovine Serum
Sterile-Filtered
Sterile-filtered
Gibco™
SDS
Documents

Documents

Promotions

Promotions