missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Bovine Serum, New Zealand origin
US-qualified performance sera for general cell culture with common cell lines
Supplier: Gibco™ 16170078
Description
- Sera Category: Secure
- Origin: New Zealand
- Collected from cattle of age 12-24 months
Cell Culture, Mammalian Cell Culture
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
New Zealand | |
Bottle | |
500 mL | |
Cattle/Bovine | |
Sustainable packaging | |
Frozen |
12 to 24 Months | |
Liquid | |
Adult Bovine Serum | |
Sterile-Filtered | |
Sterile-filtered | |
Gibco™ |