Please login to your online account to display your discounted pricing

Gibco™ BMP4 Recombinant Human Protein DFS Item


Recombinant Human Bone Morphogenetic Protein 4 (BMP4) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC9533

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHC9533

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Primary Cell Culture; Neuronal Cell Culture; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Mesenchymal Stem Cell Culture; Neural Stem Cell Culture; Differentiation & Development
Gene Alias BMP-4
Quantity 1mg
Storage Requirements Store at -5° to -30°C.
Source Human
Structural Form Recombinant/Ligand
Protein Family Growth Factors & Receptors
Expression System Mammalian
Validated Application Cell Culture
Protein Subtype BMPs (Bone Morphogenetic Proteins) & Related Factors

BMP4 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to induce alkaline phosphatase production by ATDC-5 cells

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.