missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Antibiotic-Antimycotic (100X) DFS Item

Solution contains penicillin, streptomycin, and Gibco Amphotericin B to prevent cell culture contamination.

Supplier:  Gibco™ 15240096

 View more versions of this product

Catalog No. 15240096



Description

Description

  • Gibco™ Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination
  • Solution contains 10,000 units/mL of penicillin, 10,000 μg/mL of streptomycin, and 25 μg/mL of Gibco Amphotericin B
  • The antibiotics penicillin and streptomycin prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
  • Amphotericin B prevents fungal contamination of cell cultures due to its inhibition of multi-cellular fungus and yeast
  • Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall
  • Streptomycin was originally purified from Streptomyces griseus and acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
  • Store at -5°C to -20°C
Specifications

Specifications

Storage conditions: -5 to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Mammalian Cell Culture
Antibiotic-Antimycotic
20 mL
100 X
Liquid
Sterile-filtered
Dry Ice
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.